missing translation for 'onlineSavingsMsg'
Learn More

Chloramine-T, MedChemExpress

Product Code. 16492289
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492289 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492289 Supplier Medchem Express Supplier No. HYB0959

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Chloramine-T is a titrimetric reagent, and an oxidizing agent. Chloramine-T is an oxidizing biocide.

Chloramine-T is a titrimetric reagent, and an oxidizing agent. Chloramine-T is an oxidizing biocide.

Specifications

Chemical Name or Material Chloramine-T
Physical Form Solid
CAS 127-65-1
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C7H7ClNNaO2S
Solubility Information DMSO : 130 mg/mL (571.08 mM; Need ultrasonic) ∣H2O : ≥ 41 mg/mL (180.11 mM)
SMILES O=S(C1=CC=C(C)C=C1)(N([Na])Cl)=O
Molecular Weight (g/mol) 227.64
Formula Weight 227.64
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.