missing translation for 'onlineSavingsMsg'
Learn More

Chlorambucil, TRC

Codice prodotto. 30349511
Cambia vista
Click to view available options
Quantity:
2.5 g
Dimensione della confezione:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
30349511 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30349511 Fornitore Toronto Research Chemicals N. del fornitore TRCC3250502.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorambucil
CAS 305-03-3
Molecular Formula C14 H19 Cl2 N O2
Quantity 2.5 g
Synonym Benzenebutanoic acid, 4-[bis(2-chloroethyl)amino]-, Butyric acid, 4-[p-[bis(2-chloroethyl)amino]phenyl]- (8CI), 4-[Bis(2-chloroethyl)amino]benzenebutanoic acid, 4-[p-[Bis(2-chloroethyl)amino]phenyl]butyric acid, Ambochlorin, Amboclorin, CB 1348, Chlorambucil, Chloraminophene, Chlorbutin, Chlorobutine, Ecloril, Leukeran, Leukeran Tablets, Linfolizin, Linfolysin, Lympholysin, NCI 3088, NSC 3088, γ-[p-Bis(2-chloroethyl)aminophenyl]butyric acid, γ-[p-Di(2-chloroethyl)aminophenyl]butyric acid
InChI Formula InChI=1S/C14H19Cl2NO2/c15-8-10-17(11-9-16)13-6-4-12(5-7-13)2-1-3-14(18)19/h4-7H,1-3,8-11H2,(H,18,19)
SMILES OC(=O)CCCc1ccc(cc1)N(CCCl)CCCl
IUPAC Name 4-[4-[bis(2-chloroethyl)amino]phenyl]butanoic acid
Molecular Weight (g/mol) 304.21
Formula Weight 303.0793
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.