missing translation for 'onlineSavingsMsg'
Learn More

Chloralose, TRC

Product Code. 30327819
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Product Code. Quantity unitSize
30327819 2.5 g 2.5g
1 options
This item is not returnable. View return policy

Product Code. 30327819

Brand: Toronto Research Chemicals TRCA5760052.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-Chloralose
CAS 15879-93-3
Molecular Formula C8 H11 Cl3 O6
Quantity 2.5 g
Synonym Chloralose (6CI), Chloralose, α- (8CI), α-D-Glucofuranose, 1,2-O-(2,2,2-trichloroethylidene)-, (R)-, 1,2-O-[(1R)-2,2,2-Trichloroethylidene]-α-D-glucofuranose, Furo[2,3-d]-1,3-dioxole, α-D-glucofuranose deriv., AGC, AGC (carbohydrate), Alfa 4, Alpha 4, Alphakil, Anhydroglucochloral, Aphosal, Chloralosane, Dorcalm, Dulcidor, Glucochloral, Glucochloralose, Kalmettumsomniferum, Murex, Murex (carbohydrate), Somio, α-Chloralose, α-D-Chloralose, α-Glucochloralose, α-Monotrichloroethylidene-D-glucose
InChI Formula InChI=1S/C8H11Cl3O6/c9-8(10,11)7-16-5-3(14)4(2(13)1-12)15-6(5)17-7/h2-7,12-14H,1H2/t2-,3+,4-,5-,6-,7-/m1/s1
SMILES OC[C@@H](O)[C@H]1O[C@@H]2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl
IUPAC Name (1R)-1-[(2R,3aR,5R,6S,6aR)-6-hydroxy-2-(trichloromethyl)-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol
Molecular Weight (g/mol) 309.53
Formula Weight 307.9621
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.