missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chlamydia trachomatis serovar L2 ompA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117609
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
ompA acts as a fimbriae subunit.
Specifications
| Chlamydia trachomatis serovar L2 ompA | |
| Polyclonal | |
| Unconjugated | |
| ompA | |
| Fimbrin; ompP5; Outer membrane protein P5 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 917123 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 3.66 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P0A911 | |
| ompA | |
| Recombinant Chlamydia trachomatis serovar L2 Major outer membrane porin protein (23-394aa). | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction