missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Chlamydia trachomatis Native Protein

Product Code. 18611608 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18611608 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18611608 Supplier R&D Systems™ Supplier No. NBP262417
  • ONLINE EXCLUSIVE PRICE
    £614.86
    £520.80 / 1mL
    Save £94.06
    15% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Native Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 4.9 mg/mL
For Use With (Application) Immunoassay
Formulation PBS
Quantity 1 mL
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias C.trachomatis
Research Category Virology Bacteria and Parasites
Product Line Novus
Protein Chlamydia trachomatis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.