missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chlamydia trachomatis MOMP Monoclonal Antibody (CL12-707.7), Invitrogen™

Mouse Monoclonal Antibody
£367.00 - £916.00
Specifications
| Antigen | Chlamydia trachomatis MOMP |
|---|---|
| Clone | CL12-707.7 |
| Concentration | 1.3 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17850296
|
Invitrogen™
MA5-46029 |
100 μg |
£367.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11538932
![]() |
Invitrogen™
MA110665 |
500 μg |
£480.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17800176
|
Invitrogen™
MA5-46061 |
1 mg |
£916.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MA1-10665 detects Chlamydia trachomatis MOMP from C. trachomatis serovars A, B, Ba, C, D, E, F, G, H, I, J, K, L1, L2, and L3 samples. MA1-10665 has been successfully used in ELISA applications. The MA1-10665 immunogen is C. trachomatis elementary bodies.
Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.Specifications
| Chlamydia trachomatis MOMP | |
| 1.3 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| C. trachomatis elementary bodies. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles |
| CL12-707.7 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| C. trachomatis MOMP; Chlamydia MOMP | |
| IgG2a | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title