missing translation for 'onlineSavingsMsg'
Learn More

Chlamydia Trachomatis Lps Antibody (1312/284), DyLight 650, Novus Biologicals™

Product Code. 18431094 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431094 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431094 Supplier Novus Biologicals Supplier No. NBP128820C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Chlamydia Trachomatis Lps Monoclonal antibody specifically detects Chlamydia Trachomatis Lps in Bacteria samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chlamydia Trachomatis Lps
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1312/284
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias C. trachomatis Lipopolysaccharide;Chlamydia trachomatis Lipopolysaccharide
Host Species Mouse
Immunogen Chlamydia trachomatis L2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.