missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlamydia Trachomatis Lps Antibody (1312/284), DyLight 488, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Chlamydia Trachomatis Lps Monoclonal antibody specifically detects Chlamydia Trachomatis Lps in Bacteria samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Chlamydia Trachomatis Lps |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1312/284 |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C. trachomatis Lipopolysaccharide;Chlamydia trachomatis Lipopolysaccharide |
| Host Species | Mouse |
| Immunogen | Chlamydia trachomatis L2 |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?