missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlamydia Trachomatis Lps Antibody (1312/284), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28820AF350
This item is not returnable.
View return policy
Description
Chlamydia Trachomatis Lps Monoclonal specifically detects Chlamydia Trachomatis Lps in Bacteria samples. It is validated for Western Blot, ELISA.
Specifications
| Chlamydia Trachomatis Lps | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Chlamydia trachomatis L2 | |
| Primary | |
| Bacteria | |
| IgG1 |
| Western Blot, ELISA | |
| 1312/284 | |
| Western Blot, ELISA | |
| Mouse | |
| 0.1 mL | |
| CHLAMYDIA TRACHOMATIS LPS | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction