missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlamydia Antibody (CF 6J12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64440
This item is not returnable.
View return policy
Description
Chlamydia Monoclonal specifically detects Chlamydia in Bacteria samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Chlamydia | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| NB100-64440 recognizes a genus specific epitope within Chlamydia lipopolysaccharide (LPS). LPS is a common feature of the outer envelope of gram negative bacteria, which acts as a potent endotoxin, triggering an innate immune response. Studies have shown however, that whilst the LPS of Chlamydia trachomatis does evoke an immune response, it displays only weak endotoxic activity when compared to that of other bacteria such as Salmonella Minnesota or Neisseria gonorrhoeae . | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| CF 6J12 | |
| Unconjugated | |
| Chlamydia trachomatis | |
| Elementary bodies from C. trachomatis strain SAF2. | |
| 0.5 mg | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction