missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CHL-1/L1CAM-2 Antibody Pair [HRP]

Product Code. 18229306 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18229306

Brand: Novus Biologicals™ NBP27945015PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CHL-1/L1CAM-2.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.25 pg/mL
Assay Sensitivity 31.25-2000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CHL-1/L1CAM-2.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), FLJ44930, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein
Gene ID (Entrez) 10752
Gene Symbol CHL1
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 80 ng of recombinant CHL-1/L1CAM-2.
  • Capture Antibody: 0.2 mg/mL of mouse anti-CHL-1/L1CAM-2 monoclonal antibody,
  • Detection Antibody: 0.5 mg/mL mouse anti-CHL-1/L1CAM-2 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target CHL-1/L1CAM-2
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 2 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.