Learn More
Eurofins DiscoverX CHK2 (R145W) Protein, Active
Description
Checkpoint kinase (CHK) is activated by a broad spectrum of DNA-damaging agents, including DNA-strand-breaking agents such as ionizing radiation and topoisomerase inhibitors, and those agents that cause replication stress such as ultraviolet light, hydroxyurea and 5-fluorouracil. CHK2 might have a redundant and supportive role in checkpoints, but its role in ionizing radiation-induced apoptosis is more prominent. However, its role in other DNA-damage-induced apoptosis mechanisms is less well established. CHK2 inhibitors are expected to sensitize proliferating cells to chemotherapy-induced apoptosis.
GenBank NP_009125
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 89.6 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal GST and C-terminal 6His tags |
| Expression System | Recombinant enzyme expressed in E. coli cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥50% |
| Protein | CHK2 (R145W), active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.