missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chk2 Antibody (73C175.1.1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27403F
This item is not returnable.
View return policy
Description
Chk2 Monoclonal specifically detects Chk2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Chk2 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| CHEK2 | |
| A portion of amino acids 100-150 of human Chk2 protein was used as immunogen for this antibody. | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Stem Cell Markers | |
| 11200 | |
| Store at 4C in the dark. | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 73C175.1.1 | |
| Immunohistochemistry-Paraffin | |
| CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, S.pombe) homolog | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction