missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chk2 Antibody (235303) [mFluor Violet 450 SE], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB22851MFV450
This item is not returnable.
View return policy
Description
Chk2 Monoclonal antibody specifically detects Chk2 in Mouse samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Chk2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, S.pombe) homolog | |
| E. coli-derived recombinant mouse Chk2, Met1-Ala234, Accession # Q9Z265 | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Stem Cell Markers | |
| 11200 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot | |
| 235303 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction