missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chk1 Rabbit anti-Human, Clone: 0S3Q1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16353-20UL
This item is not returnable.
View return policy
Description
Chk1 Monoclonal antibody specifically detects Chk1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Chk1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0S3Q1 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Checkpoint, S. pombe, homolog of, 1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Chk1 (O14757). | |
| 20 μg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Stem Cell Markers, Tumor Suppressors | |
| 1111 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction