missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX CHK1 Protein, Active

Product Code. 17340154
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17340154

Brand: Eurofins DiscoverX 14346

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal GST tagged, human full length CHK1, for use in Kinase Assays.

Checkpoint kinase (CHK) is activated by a broad spectrum of DNA-damaging agents, including DNA-strand-breaking agents such as ionizing radiation and topoisomerase inhibitors, and those agents that cause replication stress such as ultraviolet light, hydroxyurea and 5-fluorouracil. CHK1 regulates numerous checkpoint pathways, including S-phase and G2-M checkpoints. Other potential CHK1 functions include replication, chromatin remodelling and DNA repair. CHK1 inhibitors may sensitize tumors to chemotherapy and radiotherapy.

GenBank AF016582

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 81.6 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal GST
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade ≥70%
Protein CHK1, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.