Learn More
Eurofins DiscoverX CHK1 Protein, Active
Description
Checkpoint kinase (CHK) is activated by a broad spectrum of DNA-damaging agents, including DNA-strand-breaking agents such as ionizing radiation and topoisomerase inhibitors, and those agents that cause replication stress such as ultraviolet light, hydroxyurea and 5-fluorouracil. CHK1 regulates numerous checkpoint pathways, including S-phase and G2-M checkpoints. Other potential CHK1 functions include replication, chromatin remodelling and DNA repair. CHK1 inhibitors may sensitize tumors to chemotherapy and radiotherapy.
GenBank AF016582
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 81.6 kDa |
| Quantity | 250 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal GST |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥70% |
| Protein | CHK1, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.