missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chk1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£240.00 - £435.00
Specifications
| Antigen | Chk1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18275603
|
Novus Biologicals
NBP2-58441 |
100 μL |
£435.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678186
|
Novus Biologicals
NBP2-58441-25ul |
25 μL |
£240.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chk1 Polyclonal specifically detects Chk1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Chk1 | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Stem Cell Markers, Tumor Suppressors | |
| Checkpoint, S. pombe, homolog of, 1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1 | |
| CHEK1 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 1111 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KRREFHAEPVDVWSCGIVLTAMLAGELPWDQPSDSCQEYSDWKEKKTYLNPWKKIDSAPLALLHKILVENPSARITIPDIKKDRWYNKPLKKGAKRPRVTSGGVSESPSGFSKHIQSNLDFSPVNSASSE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title