missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chk1 Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-311H
This item is not returnable.
View return policy
Description
Chk1 Polyclonal antibody specifically detects Chk1 in Human samples. It is validated for Western Blot
Specifications
| Chk1 | |
| Polyclonal | |
| Western Blot | |
| Checkpoint, S. pombe, homolog of, 1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1 | |
| The epitope recognized by NB100-311 maps to a region between residues 250 and 300 of human Checkpoint Kinase 1 using the numbering given in entry NP_001265.1 (GeneID 1111). | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Stem Cell Markers, Tumor Suppressors | |
| 1111 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction