missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chitotriosidase Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582395
This item is not returnable.
View return policy
Description
Immunogen sequence: FDDVESFKTK VSYLKQKGLG GAMVWALDLD DFAGFSCNQG RYPLIQTLRQ ELSLPYLPSG TPELEVPKPG QPSEPEHGPS PGQDTFCQGK ADGLYPNPRE RSSFYSCAAG RLFQQSCPTG LVFSNSCKCC TWN Highest antigen sequence indentity to the following orthologs: Rat - 73%, Mouse - 70%.
Chitotriosidase is secreted by activated human macrophages and is markedly elevated in plasma of Gaucher disease patients. The expression of chitotriosidase occurs only at a late stage of differentiation of monocytes to activated macrophages in culture. Human macrophages can synthesize a functional chitotriosidase, a highly conserved enzyme with a strongly regulated expression. This enzyme may play a role in the degradation of chitin-containing pathogens.
Specifications
| Chitotriosidase | |
| Polyclonal | |
| Unconjugated | |
| CHIT1 | |
| 2300002L19Rik; CHI3; CHIT; Chit1; CHITD; chitinase 1; chitinase 1 (chitotriosidase); chitinase-1; Chitotriosidase; Chitotriosidase-1; EC 3.2.1.14; FLJ00314; MGC125322; plasma methylumbelliferyl tetra-N-acetylchitotetraoside hydrolase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1118 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13231 | |
| CHIT1 | |
| Recombinant protein corresponding to Human CHIT1. Recombinant protein control fragment (Product #RP-90229). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction