missing translation for 'onlineSavingsMsg'
Learn More

Chitinase 3-like 1 Rabbit anti-Human, Mouse, Clone: 9P1P6, Novus Biologicals™

Product Code. 18384712 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384712 20 μg 20µL
18304586 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18384712 Supplier Novus Biologicals Supplier No. NBP31607720UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Chitinase 3-like 1 Monoclonal antibody specifically detects Chitinase 3-like 1 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chitinase 3-like 1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 9P1P6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias AW208766, Brp39, Chi3l1, gp39
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 284-383 of human Chitinase 3-like 1 (NP_001267.2).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1116
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.