missing translation for 'onlineSavingsMsg'
Learn More

Chitin, MP Biomedicals™

Product Code. 11453270 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453270 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453270 Supplier MP Biomedicals Supplier No. 0210133480
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chitin is a cellulose-like biopolymer consisting predominantly of unbranched chains of b-(1->4)-2-acetamido-2-deoxy-D-glucose (also named N-acetyl-D-glucosamine) residues.

The effect of BES and other zwitterionic buffers on the Bicinchoninic acid (BCA) measurement of microgram quantities of protein has been studied. BES was found to diminish color development of the protein BCA complex by 30–35% at 50mM, and by <10% at 5mM, measuring in the 1–10mg range of BSA against protein-BCA complexes formed in water. This color loss may result from binding of Cu2+ to BES.

Other uses include:

  • A protocol has been reported for the use of BES buffered saline in the calcium phosphate mediated transfection of eukaryotic cells with plasmid DNA
  • BES may be employed as binding buffer in modified Eagle′s medium during the binding assay of human melanoma cells
  • It may be used as biobuffer to investigate the aqueous medium self-assembly of heterometallic CuII/Li 3D coordination polymers.

Specifications

CAS 1398-61-4
Melting Point >300°C (decomposition)(literature)
Molecular Formula (C8H13NO5)nC16H28N2O11
Quantity 100 g
Synonym chitin from shrimp shells, n-2r-2,4,5-trihydroxy-6-hydroxymethyl oxan-3-yl acetamide, chitin from shrimp shells, practical grade, powder, chitin from shrimp shells, vetec tm reagent grade, chitin from shrimp shells, practical grade, coarse flakes, chitin from shrimp shells, bioreagent, suitable for analysis of chitinase, purified powder
Solubility Information Soluble in concentrated HCl, H3SO4 or anhydrous Acetic Acid; practically insoluble in water, dil. acids, dil. and concentrated alkalies, alcohol and other organic solvents. There are substantial variations in solubility.
Formula Weight 770.8
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.