missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies CHIR99021

Product Code. 15672517
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15672517 1 mg 1mg
17190012 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15672517 Supplier STEMCELL Technologies Supplier No. 72052
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

WNT pathway activator; Inhibits GSK3

  • CHIR99021 is an aminopyrimidine derivative that is an extremely potent glycogen synthase kinase (GSK) 3 inhibitor, inhibiting both GSK3β (IC50 = 6.7 nM) and GSK3α (IC50 = 10 nM)
  • GSK3 is a serine/threonine kinase that is a key inhibitor of the WNT pathway; therefore CHIR99021 functions as a WNT activator
  • It shows little activity against a large panel of kinases including CDK2 and other serine/threonine kinases such as MAPK and PKB

Specifications

CAS 252917-06-9
Synonym CT 99021
Quantity 1 mg
Molecular Formula C22H18Cl2N8
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.