missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CHIR 99021 trihydrochloride
GREENER_CHOICE

Product Code. 18798537
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
18798537 10 mg 10mg
18769007 50 mg 50mg
2 options
This item is not returnable. View return policy

Product Code. 18798537

Brand: Tocris Bioscience™ 4953/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrochloride salt of CHIR 99021 (Cat. No. 4423); selective GSK-3 inhibitor

Hydrochloride salt of CHIR 99021 (Cat. No. 4423), a highly selective GSK-3 inhibitor.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1782235-14-6
Molecular Formula C22H21Cl5N8

Specifications

Quantity 10 mg
Formula Weight 574.72
Purity >99%
Molecular Weight (g/mol) 574.72
Chemical Name or Material CHIR 99021 trihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.