missing translation for 'onlineSavingsMsg'
Learn More

CHIR-98014, MedChemExpress

Product Code. 16465538
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465538 10 mg 10mg
16461707 50 mg 50mg
16438247 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16465538 Supplier Medchem Express Supplier No. HY13076
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CHIR-98014 is a potent, cell-permeable GSK-3 inhibitor with IC50s of 0.65 and 0.58 nM for GSK-3α and GSK-3β, respectively; it shows less potent activities against cdc2 and erk2.

CHIR-98014 is a potent, cell-permeable GSK-3 inhibitor with IC50s of 0.65 and 0.58 nM for GSK-3α and GSK-3β, respectively

Specifications

Chemical Name or Material CHIR-98014
CAS 252935-94-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H17Cl2N9O2
Quantity 10 mg
Solubility Information DMSO : 12.5 mg/mL (25.70 mM; ultrasonic and warming and heat to 60°C)
SMILES ClC1=CC(Cl)=CC=C1C2=NC(NCCNC3=CC=C([N+]([O-])=O)C(N)=N3)=NC=C2N4C=CN=C4
Molecular Weight (g/mol) 486.31
Formula Weight 486.31
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.