missing translation for 'onlineSavingsMsg'
Learn More

CHIPS Antibody (JCC1) - C-terminus, Novus Biologicals™

Product Code. 18392152 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392152 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392152 Supplier Novus Biologicals Supplier No. NBP311238

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CHIPS Monoclonal specifically detects CHIPS in Bacteria samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHIPS
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Immunoassay
Classification Monoclonal
Clone JCC1
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias Chemotaxis Inhibiting Protein of Staphylococcus aureus, Chemotaxis inhibitory protein, Chemotaxis-inhibiting protein, CHP, chs
Host Species Mouse
Immunogen Monoclonal antibody JCC1 reacts with the C-terminus of CHIPS.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3921863
Target Species Bacteria
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.