missing translation for 'onlineSavingsMsg'
Learn More

Manutan Chipboard shelf for tubular shelving, 1250 x 300mm

Product Code. 12845658
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12845658 Supplier Manutan Supplier No. A360396

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal for the storage of smaller items, 3mm thick. Use to cover tubular shelves with easy click-in system.

  • Thickness: 3mm
  • Depth: 300mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.