missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHIP/STUB1 Rabbit anti-Human, Clone: 34, Novus Biologicals™
Rabbit Monoclonal Antibody
£296.00 - £429.00
Specifications
| Antigen | CHIP/STUB1 |
|---|---|
| Clone | 34 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100-1:500, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1-4 ul/mg of lysate, Immunofluorescence |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18257477
|
Novus Biologicals
NBP2-90116-50UL |
50 μg |
£296.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18274718
|
Novus Biologicals
NBP2-90116-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHIP/STUB1 Monoclonal specifically detects CHIP/STUB1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| CHIP/STUB1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100-1:500, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1-4 ul/mg of lysate, Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Antigen NY-CO-7, Carboxy terminus of Hsp70-interacting protein, CHIPSTIP1 homology and U box-containing protein 1, CLL-associated antigen KW-8, E3 ubiquitin-protein ligase CHIP, EC 6.3.2.-, heat shock protein A binding protein 2 (c-terminal), HSPABP2, NY-CO-7, SDCCAG7, serologically defined colon cancer antigen 7, STIP1 homology and U-box containing protein 1, STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase, UBOX1 | |
| Recombinant Human CHIP/STUB1 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 34 | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| 0.2 um filtered solution in PBS | |
| 10273 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title