missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHIP/STUB1 Antibody (034), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90116PE
This item is not returnable.
View return policy
Description
CHIP/STUB1 Monoclonal specifically detects CHIP/STUB1 in Human samples. It is validated for Flow Cytometry.
Specifications
| CHIP/STUB1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 34 | |
| Flow Cytometry | |
| Antigen NY-CO-7, Carboxy terminus of Hsp70-interacting protein, CHIPSTIP1 homology and U box-containing protein 1, CLL-associated antigen KW-8, E3 ubiquitin-protein ligase CHIP, EC 6.3.2.-, heat shock protein A binding protein 2 (c-terminal), HSPABP2, NY-CO-7, SDCCAG7, serologically defined colon cancer antigen 7, STIP1 homology and U-box containing protein 1, STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase, UBOX1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CHIP/STUB1 isoform 1 (Q9UNE7-1; Met 1-Tyr 303). | |
| 0.1 mL | |
| Neuroscience | |
| 10273 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction