missing translation for 'onlineSavingsMsg'
Learn More

Chinese Pharmacopoeia Pesticides Mixture 147 20-100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30324017
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324017 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324017 Supplier Dr. Ehrenstorfer Supplier No. DREA50000147AL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aldicarb∣Aldicarb-sulfone∣Aldicarb-sulfoxide∣Cadusafos∣Carbofuran∣Carbofuran-3-hydroxy∣Chlordimeform free base∣Chlorsulfuron∣Coumaphos∣Demeton (O+S)∣Ethametsulfuron-methyl∣Ethoprophos∣Fenamiphos∣Fenamiphos-sulfone∣Fenamiphos-sulfoxide∣Fonofos∣Isazofos∣Isocarbofos∣Isofenphos-methyl∣Methamidophos∣Metsulfuron-methyl∣Monocrotophos∣Phorate∣Phorate-sulfone∣Phorate-sulfoxide∣Phosfolan∣Phosphamidon∣Sulfotep∣Terbufos-sulfone∣Terbufos-sulfoxide
CAS 116-06-3, 1646-88-4, 1646-87-3, 95465-99-9, 1563-66-2, 16655-82-6
Analyte or Component Names Aldicarb∣Aldicarb-sulfone∣Aldicarb-sulfoxide∣Cadusafos∣Carbofuran∣Carbofuran-3-hydroxy∣Chlordimeform free base∣Chlorsulfuron∣Coumaphos∣Demeton (O+S)∣Ethametsulfuron-methyl∣Ethoprophos∣Fenamiphos∣Fenamiphos-sulfone∣Fenamiphos-sulfoxide∣Fonofos∣Isazofos∣Isocarbofos∣Isofenphos-methyl∣Methamidophos∣Metsulfuron-methyl∣Monocrotophos∣Phorate∣Phorate-sulfone∣Phorate-sulfoxide∣Phosfolan∣Phosphamidon∣Sulfotep∣Terbufos-sulfone∣Terbufos-sulfoxide
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Isazofos 20 μg/mL∣Cadusafos 40 μg/mL∣Chlordimeform free base 40 μg/mL∣Demeton (O+S) 40 μg/mL∣Ethoprophos 40 μg/mL∣Fenamiphos 40 μg/mL∣Fenamiphos-sulfone 40 μg/mL∣Fenamiphos-sulfoxide 40 μg/mL∣Fonofos 40 μg/mL∣Isofenphos-methyl 40 μg/mL∣Phorate 40 μg/mL∣Phorate-sulfone 40 μg/mL∣Phorate-sulfoxide 40 μg/mL∣Sulfotep 40 μg/mL∣Terbufos-sulfone 40 μg/mL∣Terbufos-sulfoxide 40 μg/mL∣Monocrotophos 60 μg/mL∣Phosfolan 60 μg/mL∣Aldicarb 100 μg/mL∣Aldicarb-sulfone 100 μg/mL∣Aldicarb-sulfoxide 100 μg/mL∣Carbofuran 100 μg/mL∣Carbofuran-3-hydroxy 100 μg/mL∣Chlorsulfuron 100 μg/mL∣Coumaphos 100 μg/mL∣Ethametsulfuron-methyl 100 μg/mL∣Isocarbofos 100 μg/mL∣Methamidophos 100 μg/mL∣Metsulfuron-methyl 100 μg/mL∣Phosphamidon 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.