missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chinese Pharmacopoeia Pesticides Mixture 146 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Aldicarb∣Aldicarb-sulfone∣Aldicarb-sulfoxide∣Cadusafos∣Carbofuran∣Carbofuran-3-hydroxy∣Chlordimeform free base∣Chlorsulfuron∣Coumaphos∣Demeton (O+S)∣Ethametsulfuron-methyl∣Ethoprophos∣Fenamiphos∣Fenamiphos-sulfone∣Fenamiphos-sulfoxide∣Fonofos∣Isazofos∣Isocarbofos∣Isofenphos-methyl∣Methamidophos∣Metsulfuron-methyl∣Monocrotophos∣Phorate∣Phorate-sulfone∣Phorate-sulfoxide∣Phosfolan∣Phosphamidon∣Sulfotep∣Terbufos-sulfone∣Terbufos-sulfoxide |
| CAS | 116-06-3, 1646-88-4, 1646-87-3, 95465-99-9, 1563-66-2, 16655-82-6 |
| Analyte or Component Names | Aldicarb∣Aldicarb-sulfone∣Aldicarb-sulfoxide∣Cadusafos∣Carbofuran∣Carbofuran-3-hydroxy∣Chlordimeform free base∣Chlorsulfuron∣Coumaphos∣Demeton (O+S)∣Ethametsulfuron-methyl∣Ethoprophos∣Fenamiphos∣Fenamiphos-sulfone∣Fenamiphos-sulfoxide∣Fonofos∣Isazofos∣Isocarbofos∣Isofenphos-methyl∣Methamidophos∣Metsulfuron-methyl∣Monocrotophos∣Phorate∣Phorate-sulfone∣Phorate-sulfoxide∣Phosfolan∣Phosphamidon∣Sulfotep∣Terbufos-sulfone∣Terbufos-sulfoxide |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Aldicarb 100 μg/mL∣Aldicarb-sulfone 100 μg/mL∣Aldicarb-sulfoxide 100 μg/mL∣Cadusafos 100 μg/mL∣Carbofuran 100 μg/mL∣Carbofuran-3-hydroxy 100 μg/mL∣Chlordimeform free base 100 μg/mL∣Chlorsulfuron 100 μg/mL∣Coumaphos 100 μg/mL∣Demeton (O+S) 100 μg/mL∣Ethametsulfuron-methyl 100 μg/mL∣Ethoprophos 100 μg/mL∣Fenamiphos 100 μg/mL∣Fenamiphos-sulfone 100 μg/mL∣Fenamiphos-sulfoxide 100 μg/mL∣Fonofos 100 μg/mL∣Isazofos 100 μg/mL∣Isocarbofos 100 μg/mL∣Isofenphos-methyl 100 μg/mL∣Methamidophos 100 μg/mL∣Metsulfuron-methyl 100 μg/mL∣Monocrotophos 100 μg/mL∣Phorate 100 μg/mL∣Phorate-sulfone 100 μg/mL∣Phorate-sulfoxide 100 μg/mL∣Phosfolan 100 μg/mL∣Phosphamidon 100 μg/mL∣Sulfotep 100 μg/mL∣Terbufos-sulfone 100 μg/mL∣Terbufos-sulfoxide 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?