missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chinese Pharmacopoeia Pesticides Mixture 145 40-100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Chlordimeform free base∣Coumaphos∣delta-HCH∣Demeton (O+S)∣Dicofol∣Dieldrin∣Endosulfan-sulfate∣Ethoprophos∣Fenamiphos∣Fipronil∣Fipronil Sulfide∣Fipronil Sulfone∣Fipronil-desulfinyl∣gamma-HCH∣Isocarbofos∣Isofenphos-methyl∣Monocrotophos∣Nitrofen∣Parathion-ethyl∣Parathion-methyl∣Phorate∣Phospholan-methyl∣Sulfotep∣Terbufos |
| CAS | 789-02-6, 72-54-8, 72-55-9, 50-29-3, 309-00-2, 959-98-8 |
| Analyte or Component Names | 2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Chlordimeform free base∣Coumaphos∣delta-HCH∣Demeton (O+S)∣Dicofol∣Dieldrin∣Endosulfan-sulfate∣Ethoprophos∣Fenamiphos∣Fipronil∣Fipronil Sulfide∣Fipronil Sulfone∣Fipronil-desulfinyl∣gamma-HCH∣Isocarbofos∣Isofenphos-methyl∣Monocrotophos∣Nitrofen∣Parathion-ethyl∣Parathion-methyl∣Phorate∣Phospholan-methyl∣Sulfotep∣Terbufos |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Chlordimeform free base 40 μg/mL∣Demeton (O+S) 40 μg/mL∣Ethoprophos 40 μg/mL∣Fenamiphos 40 μg/mL∣Fipronil 40 μg/mL∣Fipronil Sulfide 40 μg/mL∣Fipronil Sulfone 40 μg/mL∣Fipronil-desulfinyl 40 μg/mL∣Isofenphos-methyl 40 μg/mL∣Parathion-ethyl 40 μg/mL∣Parathion-methyl 40 μg/mL∣Phorate 40 μg/mL∣Sulfotep 40 μg/mL∣Terbufos 40 μg/mL∣Monocrotophos 60 μg/mL∣Phospholan-methyl 60 μg/mL∣2,4'-DDT 100 μg/mL∣4,4'-DDD 100 μg/mL∣4,4'-DDE 100 μg/mL∣4,4'-DDT 100 μg/mL∣Aldrin 100 μg/mL∣alpha-Endosulfan 100 μg/mL∣alpha-HCH 100 μg/mL∣beta-Endosulfan 100 μg/mL∣beta-HCH 100 μg/mL∣Coumaphos 100 μg/mL∣delta-HCH 100 μg/mL∣Dicofol 100 μg/mL∣Dieldrin 100 μg/mL∣Endosulfan-sulfate 100 μg/mL∣gamma-HCH 100 μg/mL∣Isocarbofos 100 μg/mL∣Nitrofen 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?