missing translation for 'onlineSavingsMsg'
Learn More

Chinese Pharmacopoeia Pesticides Mixture 144 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30323281
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323281 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323281 Supplier Dr. Ehrenstorfer Supplier No. DREA50000144AL
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Chlordimeform free base∣Coumaphos∣delta-HCH∣Demeton (O+S)∣Dicofol∣Dieldrin∣Endosulfan-sulfate∣Ethoprophos∣Fenamiphos∣Fipronil∣Fipronil Sulfide∣Fipronil Sulfone∣Fipronil-desulfinyl∣gamma-HCH∣Isocarbofos∣Isofenphos-methyl∣Monocrotophos∣Nitrofen∣Parathion-ethyl∣Parathion-methyl∣Phorate∣Phospholan-methyl∣Sulfotep∣Terbufos
CAS 789-02-6, 72-54-8, 72-55-9, 50-29-3, 309-00-2, 959-98-8
Analyte or Component Names 2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Chlordimeform free base∣Coumaphos∣delta-HCH∣Demeton (O+S)∣Dicofol∣Dieldrin∣Endosulfan-sulfate∣Ethoprophos∣Fenamiphos∣Fipronil∣Fipronil Sulfide∣Fipronil Sulfone∣Fipronil-desulfinyl∣gamma-HCH∣Isocarbofos∣Isofenphos-methyl∣Monocrotophos∣Nitrofen∣Parathion-ethyl∣Parathion-methyl∣Phorate∣Phospholan-methyl∣Sulfotep∣Terbufos
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2,4'-DDT 100 μg/mL∣4,4'-DDD 100 μg/mL∣4,4'-DDE 100 μg/mL∣4,4'-DDT 100 μg/mL∣Aldrin 100 μg/mL∣alpha-Endosulfan 100 μg/mL∣alpha-HCH 100 μg/mL∣beta-Endosulfan 100 μg/mL∣beta-HCH 100 μg/mL∣Chlordimeform free base 100 μg/mL∣Coumaphos 100 μg/mL∣delta-HCH 100 μg/mL∣Demeton (O+S) 100 μg/mL∣Dicofol 100 μg/mL∣Dieldrin 100 μg/mL∣Endosulfan-sulfate 100 μg/mL∣Ethoprophos 100 μg/mL∣Fenamiphos 100 μg/mL∣Fipronil 100 μg/mL∣Fipronil Sulfide 100 μg/mL∣Fipronil Sulfone 100 μg/mL∣Fipronil-desulfinyl 100 μg/mL∣gamma-HCH 100 μg/mL∣Isocarbofos 100 μg/mL∣Isofenphos-methyl 100 μg/mL∣Monocrotophos 100 μg/mL∣Nitrofen 100 μg/mL∣Parathion-ethyl 100 μg/mL∣Parathion-methyl 100 μg/mL∣Phorate 100 μg/mL∣Phospholan-methyl 100 μg/mL∣Sulfotep 100 μg/mL∣Terbufos 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.