missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chinese Hamster PLBD2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£161.00 - £461.00
Specifications
| Antigen | PLBD2 |
|---|---|
| Clone | 2751B |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18756255
|
R&D Systems
MAB106921 |
100 μg |
£461.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18122271
|
R&D Systems
MAB106921-100 |
100 μg |
£444.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18746215
![]() |
R&D Systems
MAB106921-SP |
25 μg |
£161.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLBD2 Monoclonal antibody specifically detects PLBD2 in Chinese Hamster samples. It is validated for ELISASpecifications
| PLBD2 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| LAMA-like protein 2,76 kDa protein, lamina ancestor homolog 2, mannose-6-phosphate protein associated protein p76, P76, phospholipase B domain containing 2, phospholipase B domain-containing protein 2, phospholipase B-like 2 32 kDa form, phospholipase B-like 2 45 kDa form, PLB homolog 2, putative phospholipase B-like 2 | |
| Recombinant chinese hamster PLBD-2, Met1-Asp585, Accession # XP_003510860 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2751B | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Chinese Hamster | |
| XP_003510860 | |
| 196463 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title