missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chimaerin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | Chimaerin 2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Chimaerin 2 Polyclonal specifically detects Chimaerin 2 in Human samples. It is validated for Western Blot.Specifications
| Chimaerin 2 | |
| Polyclonal | |
| Rabbit | |
| P52757 | |
| 1124 | |
| IgG | |
| 38 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| ARHGAP3beta chimerin, BCH, beta-chimaerin, beta-chimerin, chimerin (chimaerin) 2, MGC138360, Rho GTPase-activating protein 3, RhoGAP3, rho-GTPase-activating protein 3 | |
| CHN2 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title