missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chimaerin 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92581-0.1ml
This item is not returnable.
View return policy
Description
Chimaerin 2 Polyclonal antibody specifically detects Chimaerin 2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Chimaerin 2 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000 | |
| ARHGAP3beta chimerin, BCH, beta-chimaerin, beta-chimerin, chimerin (chimaerin) 2, MGC138360, Rho GTPase-activating protein 3, RhoGAP3, rho-GTPase-activating protein 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human CHN2 (NP_004058.1). MAASSNSSLSGSSVSSDAEEYQPPIWKSYLYQLQQEAPRPKRIICPREVENRPKYYGREFHGIISREQADELLGGVEGAYILRESQRQPGCYTLALRFGNQTLNYRLFHDGKHFVGEKRFESIHDLVTDGLITLYIETKAAEYISKMTTNPIYEHIGYATLLREKVSRRLSRSKNEPRKTNVTHEEHTAVEKISSLVRRA | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 1124 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction