Learn More
Thermo Scientific™ Chiller Plumbing Kits and Accessories
Product Code. 17108165 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Fittings
Plumbing Package
Tubing
Tubing Kit
For Use With (Equipment):
Merlin Chillers with PD1/PD2 pump
ThermoFlex 3500 to 5000
ThermoFlex 900 to 10000
ThermoTemp, Polar
ThermoTemp, Polar, VersaCool
Size:
1/2 in. NPT, 25 ft.
1/2 in. NPT, 50 ft.
1/2 to 3/8 in. NPT
3/4 in. NPT, 32.8 ft.
6 mm ID per meter, 0.5 meter long
6 mm ID per meter, 1.0 m
6 mm ID per meter, 1.0 meter long
6 mm ID per meter, 1.5 m
6 mm ID per meter, 1.5 meter long
8 mm ID per meter
Material:
Brass
Flexible Plastic
Perbunan
Polybraid
Stainless-steel
Unit Size:
1m
Each
Pack of 2
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Size Material unitSize
17108165 Plumbing Package ThermoFlex 3500 to 5000 3/4 in. NPT, 32.8 ft. Polybraid Each
15176445 Plumbing Package Merlin Chillers with PD1/PD2 pump 1/2 in. NPT, 25 ft. Flexible Plastic Each
11957514 Tubing Kit ThermoFlex 900 to 10000 1/2 in. NPT, 50 ft. Polybraid Each
10430463 Tubing ThermoTemp, Polar, VersaCool 6 mm ID per meter, 1.0 meter long Stainless-steel Each
10505872 Tubing ThermoTemp, Polar 6 mm ID per meter, 1.0 m Stainless-steel Each
10771993 Tubing ThermoTemp, Polar 6 mm ID per meter, 1.5 m Stainless-steel Each
10719181 Tubing ThermoTemp, Polar, VersaCool 6 mm ID per meter, 0.5 meter long Stainless-steel Each
10111031 Tubing ThermoTemp, Polar, VersaCool 6 mm ID per meter, 1.5 meter long Stainless-steel Each
10258621 Tubing ThermoTemp, Polar 8 mm ID per meter Perbunan 1m
15865038 Fittings ThermoFlex 900 to 10000 1/2 to 3/8 in. NPT Brass Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 17108165 Supplier Thermo Scientific™ Supplier No. 611000000107
Choose these chiller plumbing kits and accessories, which include flexible tubing, tubing insulation and hose clamps.
Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.
Features:
  • Include flexible tubing, tubing insulation, and hose clamps
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ThermoFlex 3500 to 5000
Type Plumbing Package
Includes 32.8 ft. (10 meters) of 5/8 in. ID Polybraid hose (use with 3/4 in. hose barbs) , insulation and clamps
Material Polybraid
Size 3/4 in. NPT, 32.8 ft.