Learn More
Thermo Scientific™ Chiller Plumbing Kits and Accessories
Choose these chiller plumbing kits and accessories, which include flexible tubing, tubing insulation and hose clamps.
Brand: Thermo Scientific™ 598000000011
Code : NEW
Additional Details : Weight : 0.05000kg
Description
Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.
Features:
- Include flexible tubing, tubing insulation, and hose clamps
Specifications
Hose Barbs | |
Fittings | |
1/2 to 3/8 in. NPT |
ThermoFlex™ 900 to 10000 with P1, P2 or T1 pumps | |
Two 1/2 in. MPT to 3/8 in. ID brass hose barbs |