missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHIL3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA581356

Product Code. 16328384

  • £289.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse Chitinase-like 3/Chi3l3.

The CHIL3 gene encodes the protein chitinase-like 3, which is primarily secreted by macrophages and is involved in immune regulation, tissue remodeling, and inflammatory responses. CHIL3, also known as Ym1, belongs to the chitinase-like protein family but lacks chitinase activity due to mutations in its catalytic domain. It is predominantly expressed in lung tissue, where it plays a vital role in asthmatic responses and allergic inflammation. Furthermore, CHIL3 exerts anti-inflammatory roles during pathogen infection by modulating macrophage pro-inflammatory activity and promoting the Th2 immune response. Beyond immunological involvement, CHIL3 is integral to tissue repair mechanisms and cancer cell proliferation, contributing to extracellular matrix restructuring and angiogenesis. It facilitates macrophage polarization towards an M2 phenotype, which is associated with tumor progression and metastasis. Thus, CHIL3 is considered a potential biomarker for inflammatory diseases and cancer, with ongoing research examining its therapeutic modulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CHIL3
Polyclonal
Unconjugated
Chil3
AI505981; beta-N-acetylhexosaminidase Ym1; Chi3l3; Chil3; chitinase 3-like 3; chitinase-3-like protein 3; chitinase-like 3; chitinase-like protein 3; ECF-L; eosinophil chemotactic cytokine; eosinophil chemotactic factor-L; secreted protein Ym1; Ym1
Rabbit
Antigen affinity chromatography, Protein A
RUO
12655
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Western Blot, Immunocytochemistry
1 mg/mL
PBS with no preservative
O35744
Chil3
Recombinant Mouse Chitinase-like 3/Chi3l3 Protein (Met1-Tyr398).
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.