missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chikungunya Virus VLP Mouse anti-Virus, Clone: EB6-2-B6-F5, Novus Biologicals™
Mouse Monoclonal Antibody
£447.00 - £1145.00
Specifications
| Antigen | Chikungunya Virus VLP |
|---|---|
| Clone | EB6-2-B6-F5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18336746
|
Novus Biologicals
NBP3-14853-0.1MG |
0.1 mg |
£447.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18392093
|
Novus Biologicals
NBP3-14853-0.5MG |
0.5 mg |
£1145.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chikungunya Virus VLP Monoclonal antibody specifically detects Chikungunya Virus VLP in Virus samples. It is validated for Western Blot, ELISASpecifications
| Chikungunya Virus VLP | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Virus | |
| An equal mix of Chikungunya, Mayaro and O'nyong'nyong VLPs (comprising E1, E2 and capsid proteins), produced in HEK293 cells | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| EB6-2-B6-F5 | |
| Western Blot, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS pH 7.4 | |
| IgG | |
| Protein G purified | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title