missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chikungunya Virus VLP Mouse anti-Virus, Clone: AG7-2-A7-E10, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Chikungunya Virus VLP Monoclonal antibody specifically detects Chikungunya Virus VLP in Virus samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Chikungunya Virus VLP |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | AG7-2-A7-E10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA |
| Formulation | PBS pH 7.4 |
| Host Species | Mouse |
| Immunogen | An equal mix of Chikungunya, Mayaro and O'nyong'nyong VLPs (comprising E1, E2 and capsid proteins), produced in HEK293 cells |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?