missing translation for 'onlineSavingsMsg'
Learn More

Chikungunya Virus VLP Antibody, Novus Biologicals™

Product Code. 18341672 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341672 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341672 Supplier Novus Biologicals Supplier No. NBP314836

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Chikungunya Virus VLP Polyclonal antibody specifically detects Chikungunya Virus VLP in Virus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chikungunya Virus VLP
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation PBS pH 7.4
Host Species Sheep
Immunogen Recombinant CHIKV VLPs, expressed in mammalian cells
Purification Method Protein G purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.