missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHID1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | CHID1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CHID1 Polyclonal antibody specifically detects CHID1 in Mouse samples. It is validated for Western BlotSpecifications
| CHID1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS with 50% glycerol, pH7.3. | |
| 66005 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| chitinase domain containing 1, chitinase domain-containing protein 1, FLJ42707, MGC3234, SI-CLP, stabilin-1 interacting chitinase-like protein, Stabilin-1-interacting chitinase-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-393 of human CHID1 (NP_076436.3). MRTLFNLLWLALACSPVHTTLSKSDAKKAASKTLLEKSQFSDKPVQDRGLVVTDLKAESVVLEHRSYCSAKARDRHFAGDVLGYVTPWNSHGYDVTKVFGSKFTQISPVWLQLKRRGREMFEVTGLHDVDQGWMRAVRKHAKGLHIVPRLLFEDWTYDDFRNVLDSEDEIEELSKTVVQVAKNQHFDGFVVEVWNQLLSQKRVGLIHMLTHLAEALHQARLLALLVIPPAITPGTDQLGMFTHKEFEQLAPVLDGFSLMTYDYSTAHQPGPNAPLSWVRACVQVLDPKSKWRSKILLGLNFYGMDYATSKDAREPVVGARYIQTLKDHRPRMVWDSQASEHFFEYKKSRSGRHVVFYPTLKSLQVRLELARELGVGVSIWELGQGLDYFYDLL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title