missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken Netrin-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF128-SP
This item is not returnable.
View return policy
Description
Netrin-1 Polyclonal specifically detects Netrin-1 in Chicken samples. It is validated for Western Blot.
Specifications
| Netrin-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| netrin 1, netrin-1, Netrin1, NTN1, NTN1Lnetrin 1, mouse, homolog of | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9423 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q90922 | |
| NTN1 | |
| Mouse myeloma cell line NS0-derived recombinant chicken Netrin-1 (R&D Systems, Catalog # 128-N1) Gly26-Ala606 Accession # Q90922 | |
| 25 μg | |
| Primary | |
| Detects chicken Netrin-1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant chicken Netrin-2 and recombinant mouse Netrin-1 is observed. | |
| Chicken | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction