missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZGPAT, Chicken, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against synthetic peptide of ZGPAT.
Brand: Abnova PAB25752.100ug
This item is not returnable.
View return policy
Specifications
| ZGPAT | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| ZGPAT | |
| ZGPAT | |
| A synthetic peptide corresponding to 15 amino acids at C-terminus of human ZGPAT. | |
| 100 μg | |
| Primary | |
| Multiple isoforms of ZGPAT are known to exist. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgY |
| ELISA, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Chicken polyclonal antibody raised against synthetic peptide of ZGPAT. | |
| In PBS (0.02% sodium azide) | |
| GPATC6/GPATCH6/KIAA1847/MGC44880/ZC3H9/ZC3HDC9 | |
| Chicken | |
| Peptide affinity purification | |
| RUO | |
| 84619 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction