missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken anti-Rat IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB120-6836
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:1000 - 1:10000, ELISA 1:10000 - 1:100000, Immunohistochemistry 1:500 - 1:2500 | |
| Rat IgG whole molecule | |
| 0.5 mg | |
| Adaptive Immunity, Immunology | |
| Prepared from monospecific antiserum by immunoaffinity chromatography using Rat IgG coupled to agarose. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Chicken Serum, Rat IgG and Rat Serum. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| Chicken | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction