missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken anti-Rabbit IgG, HRP, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB29110.1mL
This item is not returnable.
View return policy
Specifications
| Chicken anti-Rabbit IgG | |
| Polyclonal | |
| Chicken polyclonal antibody raised against Rabbit IgG. The antibody is conjugated with HRP. | |
| In 50mM HEPES, pH 7.0, 0.1M NaCl, (0.2% BSA, 0.01% thimerosal). | |
| Purified rabbit IgG (whole protein). | |
| 1 mL | |
| Rabbit | |
| Liquid |
| ELISA, Western Blot | |
| HRP | |
| ELISA Western Blot(1:5000) The optimal working dilution should be determined by the end user. | |
| Chicken | |
| Antigen affinity purification | |
| Secondary | |
| Store at 4°C and avoid from light. Do not freeze unless for long term storage, store at -20°C with 50% glycerol. Aliquot to avoid repeated freezing and thawing. | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction