missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPZ, Chicken, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against recombinant Human MPZ.
Brand: Abnova PAB29090.200ug
This item is not returnable.
View return policy
Specifications
| MPZ | |
| Polyclonal | |
| Chicken polyclonal antibody raised against recombinant Human MPZ. | |
| In PBS, pH 7.2 (0.02% sodium azide) | |
| MPZ | |
| Two different KLH-conjugated synthetic peptides corresponding to different regions of the MPZ gene product, between the human (NP_000521, NCBI) and mouse (NP_032649, NCBI) sequences. | |
| 200 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry | |
| Unconjugated | |
| Immunocytochemistry(1:1000-1:2000) Immunohistochemistry(1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| CHM/CMT1/CMT1B/CMT2I/CMT2J/CMT4E/CMTDI3/DSS/HMSNIB/MPP/P0 | |
| Chicken | |
| Antigen affinity purification | |
| RUO | |
| 4359/17528 | |
| Store at 4°C and avoid from light. Aliquot to avoid repeated freezing and thawing. |
|
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction