missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken anti-Mouse IgG (H+L) Secondary Antibody, Texas Red, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB7598
209.95 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Texas Red | |
| Chicken | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Mouse IgG whole molecule | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| Prepared from monospecific antiserum by immunoaffinity chromatography using Mouse IgG coupled to agarose beads. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Chicken Serum, Mouse IgG and Mouse Serum. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction