missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken anti-Mouse IgG (H+L) Secondary Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
£86.00
Specifications
| Antigen | IgG (H+L) |
|---|---|
| Concentration | 4.5 mg/mL |
| Applications | Immunoassay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay.Specifications
| IgG (H+L) | |
| Immunoassay | |
| Unconjugated | |
| RUO | |
| Mouse | |
| IgG | |
| Affinity purified | |
| Based on IEP, this antibody reacts with heavy gamma chains on mouse IgG and light chains on all mouse immunoglobulins |
| 4.5 mg/mL | |
| Polyclonal | |
| Chicken | |
| Adaptive Immunity, Immunology | |
| Purified Mouse IgG (H&L) | |
| Secondary | |
| Store at 4°C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title